Skip to Content
MilliporeSigma
All Photos(1)

Documents

594806

Sigma-Aldrich

Butylboronic acid N,N,N′,N′-tetramethyl-D-tartaric acid diamide ester

97%

Synonym(s):

(4S,5S)-2-Butyl-N,N,N′N′-tetramethyl-1,3,2-dioxaborolane-4,5-dicarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H23BN2O4
CAS Number:
Molecular Weight:
270.13
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

optical activity

[α]20/D +106°, c = 1% in chloroform

refractive index

n20/D 1.4780 (lit.)

bp

305-306 °C (lit.)

density

1.096 g/mL at 25 °C (lit.)

SMILES string

CCCCB1O[C@@H]([C@H](O1)C(=O)N(C)C)C(=O)N(C)C

InChI

1S/C12H23BN2O4/c1-6-7-8-13-18-9(11(16)14(2)3)10(19-13)12(17)15(4)5/h9-10H,6-8H2,1-5H3/t9-,10-/m0/s1

InChI key

AFQWQRBBIZKYTE-UWVGGRQHSA-N

Application

Butylboronic acid N,N,N′,N′-tetramethyl-D-tartaric acid diamide is a dioxaborolane ligand that can be used to prepare:
  • Enantioenriched spiropentanes via zinc catalyzed cyclopropanation of corresponding hydroxymethylallenes.
  • 1,2,3-trisubstituted cyclopropane derivatives by asymmetric cyclopropanation of allylic alcohols in the presence of zinc reagents.
  • Glycolipids plakosides A, B, and their derivatives by Sharpless asymmetric dihydroxylation and cyclopropanation reaction.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Enantioselective synthesis of 1, 2, 3-trisubstituted cyclopropanes using gem-dizinc reagents
Zimmer LE and Charette AB
Journal of the American Chemical Society, 131(43), 15624-15626 (2009)
Total synthesis and biological evaluation of glycolipids plakosides A, B and their analogs
Nicolaou KC, et al.
Helvetica Chimica Acta, 83(8), 1977-2006 (2000)
Enantioselective synthesis of spiropentanes from hydroxymethylallenes
Charette AB, et al.
Organic Letters, 3(21), 3293-3295 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service