Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

593591

Sigma-Aldrich

(1-Chloro-2-formylvinyl)ferrocene

Synonym(s):

(1-Chloro-3-oxo-1-propenyl)ferrocene, 3-Ferrocenyl-3-chloropropenal, [(1-Chloro-2-formylvinyl)cyclopentadienyl]cyclopentadienyliron, ß-Ferrocenyl-ß-chloroacrolein

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H11ClFeO
CAS Number:
Molecular Weight:
274.52
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

solid

reaction suitability

core: iron
reagent type: catalyst

mp

74-78 °C (lit.)

SMILES string

[Fe].[CH]1[CH][CH][CH][CH]1.ClC(=CC=O)[C]2[CH][CH][CH][CH]2

InChI

1S/C8H6ClO.C5H5.Fe/c9-8(5-6-10)7-3-1-2-4-7;1-2-4-5-3-1;/h1-6H;1-5H;

InChI key

RQRKVVWRCJBKBO-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Ferrocene-based hydrazones and pyrazoles
  • Complexes with linked ferrocene moieties

Reactant for:
  • Converstion of mathyl ketones into terminal acetylenes
  • Addition and cycloaddition reaction kinetics of substituted ferrocenylketenes
  • Reaction with phenylenediamine in the presence of nickel diacetate

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service