Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

593095

Sigma-Aldrich

Diethyl (2-methylallyl)phosphonate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2POCH2C(=CH2)CH3
CAS Number:
Molecular Weight:
192.19
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.4380 (lit.)

bp

62 °C/0.1 mmHg (lit.)

density

1.013 g/mL at 25 °C (lit.)

functional group

phosphonate

SMILES string

CCOP(=O)(CC(C)=C)OCC

InChI

1S/C8H17O3P/c1-5-10-12(9,11-6-2)7-8(3)4/h3,5-7H2,1-2,4H3

InChI key

QOZGSMHGXZMADD-UHFFFAOYSA-N

General description

Diethyl (2-methylallyl)phosphonate is an organophosphorous compound.[1] It participates in the synthesis of α-aminophosphonate derivatives and azaphosphones. The analgesic/antiinflammatory properties of these derivatives were evaluated.[2]

Application

Diethyl (2-methylallyl)phosphonate can be used as a reagent in the Horner-Wadsworth-Emmons reaction to form conjugated carbon–carbon double bonds.[3]
It can also be used as a reactant for:
  • Enantioselective total synthesis of 10-isocyano-4-cadinene as antifouling agent.[4]
  • Regiospecific preparation of 4-oxo-2-alkenylphosphonates (OAP) via silylation followed by Friedel-Crafts acylation and isomerization. OAP can serve as building blocks for the construction of polyethylenic chains.[5]
  • The synthesis of azaphosphone as a potent analgesic/anti-inflammatory agents.[2]

Reactant for:
  • Enantioselective synthesis of 10-isocyano-4-cadinene and its stereoisomers with antifouling activity
  • Preparation of 4-Oxo-2-alkenylphosphonates via silylation followed by regiospecific Friedel-Crafts acylation and isomerization

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Carbodiimides-key mediators in the synthesis of novel cytotoxic and analgesic/antiinflammatory motifs based on a-amino-, enaminophosphonates, and azaphosphones.
Abdou WM, et al.
Royal Society of Chemistry Advances, 3(5), 1528-1540 (2013)
I N Smirnova et al.
The journal of physical chemistry. B, 114(50), 16936-16947 (2010-12-01)
The high brilliance of the AILES beamline at the SOLEIL synchrotron facility has been exploited for the study of the gas-phase vibrational spectra of weakly volatile organophosphorous compounds. The propagation of the synchrotron radiation in long path length gas cells
Keisuke Nishikawa et al.
Organic letters, 12(5), 904-907 (2010-02-06)
The first enantioselective total synthesis of 10-isocyano-4-cadinene, a marine sesquiterpene isolated from nudibranchs of the family Phyllidiidae, was achieved. The cadinene is expected to be a novel nontoxic antifouling agent. In the synthesis, an intermolecular Diels-Alder reaction and a SmI(2)-induced
Lee et al.
The Journal of organic chemistry, 65(13), 4175-4178 (2000-06-24)
Treatment of allylic and vinylic phosphonates with excess LiHMDS, followed by addition of chlorotrimethylsilane, afforded alpha- and gamma- silylated allylic phosphonate mixtures. Without separation, these mixtures underwent the Friedel-Crafts reaction and base-promoted isomerization to give 4-oxo-2-alkenylphosphonates, which can serve as
(-)-Sparteine-mediated stereoselective intramolecular conjugate addition reactions of dienes and enynes
Oestreich M and Hoppe D
Tetrahedron Letters, 40(10), 1881-1884 (1999)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service