Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

593036

Sigma-Aldrich

N-(4-Chlorophenyl)formamide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NHCHO
CAS Number:
Molecular Weight:
155.58
MDL number:
UNSPSC Code:
12352100

Assay

97%

mp

101-105 °C (lit.)

SMILES string

Clc1ccc(NC=O)cc1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Yoshida et al.
Sangyo igaku. Japanese journal of industrial health, 34(1), 3-9 (1992-01-01)
Previously, we identified by gas chromatography/mass spectrometry (GC/MS) urinary metabolites of p-chloroaniline (p-CA) in a patient with acute p-CA poisoning. Among these metabolites there is a possibility that 2,4-dichloroaniline (2,4-DCA) and p-chloroformanilide (p-CFA) are produced through some unknown metabolic pathways

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service