Skip to Content
MilliporeSigma
All Photos(3)

Documents

592781

Sigma-Aldrich

Ethoxynonafluorobutane

99.0%, mixture of two isomers

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5F9O
Molecular Weight:
264.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99.0%

autoignition temp.

707 °F

impurities

<100 ppm water

evapn. residue

<0.0003%

refractive index

n20/D 1.282 (lit.)

bp

76 °C (lit.)

mp

−138 °C (lit.)

density

1.428 g/mL at 25 °C (lit.)

UV absorption

λ: 220 nm Amax: 1.0
λ: 225 nm Amax: 0.35
λ: 250 nm Amax: 0.05
λ: 300-400 nm Amax: 0.01

SMILES string

CCOC(F)(F)C(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C6H5F9O/c1-2-16-6(14,15)4(9,10)3(7,8)5(11,12)13/h2H2,1H3

InChI key

DFUYAWQUODQGFF-UHFFFAOYSA-N

General description

Ethoxynonafluorobutane can be called as an eco-friendly solvent, which finds applications in chromatography since it has high lipophilic character as a result of which, it can be used as a substitute for hexane.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?Retention of fluorinated chiral selectors in biphasic fluorinated solvent systems and its application to the separation of enantiomers by countercurrent chromatography?
Perez MA and Minguillon C
Journal of Chromatography A, 1217(07), 1094-1100 (2010)
Kang Hee Ku et al.
Advanced materials (Deerfield Beach, Fla.), 31(51), e1905569-e1905569 (2019-10-23)
Dynamically reconfigurable oil-in-water (o/w) Pickering emulsions are developed, wherein the assembly of particles (i.e., platinum-on-carbon and iron-on-carbon particles) can be actively controlled by adjusting interfacial tensions. A balanced adsorption of particles and surfactants at the o/w interface allows for the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service