Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

59262

Sigma-Aldrich

2-Methoxypropene

produced by BASF, ≥98.0% (GC)

Synonym(s):

Isopropenyl methyl ether

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=C(CH3)OCH3
CAS Number:
Molecular Weight:
72.11
Beilstein:
1734635
EC Number:
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:

grade

produced by BASF

vapor pressure

27.96 psi ( 55 °C)
8.28 psi ( 20 °C)

Assay

≥98.0% (GC)

autoignition temp.

374 °F

contains

potassium carbonate as stabilizer

impurities

≤0.2% water

refractive index

n20/D 1.382 (lit.)

bp

34-36 °C (lit.)

density

0.766 g/mL at 20 °C
0.753 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

COC(C)=C

InChI

1S/C4H8O/c1-4(2)5-3/h1H2,2-3H3

InChI key

YOWQWFMSQCOSBA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the kinetic acetonation of carbohydrates

replaced by

Product No.
Description
Pricing

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Flam. Liq. 1

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

-20.2 °F - closed cup

Flash Point(C)

-29 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E. Fanton et al.
The Journal of Organic Chemistry, 46, 4057-4057 (1981)
Junya Ito et al.
Journal of chromatography. A, 1386, 53-61 (2015-02-18)
Increasing evidence suggests that phospholipid peroxidation plays important roles in the pathogenesis of various diseases, such as atherosclerosis. With regard to the biochemical processes that initiate phospholipid peroxidation in vivo, enzymatic conversion of phosphatidylcholine to phosphatidylcholine hydroperoxide (PCOOH) by lipoxygenase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service