Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

592285

Sigma-Aldrich

Tricyclohexylphosphine solution

1 M in toluene

Synonym(s):

NSC 158657, PCy3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H11)3P
CAS Number:
Molecular Weight:
280.43
Beilstein:
957171
MDL number:
UNSPSC Code:
12350000
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
reaction type: Cross Couplings

concentration

1 M in toluene

density

0.901 g/mL at 25 °C

functional group

phosphine

SMILES string

C1CCC(CC1)P(C2CCCCC2)C3CCCCC3

InChI

1S/C18H33P/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18/h16-18H,1-15H2

InChI key

WLPUWLXVBWGYMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for:
  • Carbonylative Suzuki-coupling of benzyl halides with potassium aryltrifluoroborates
  • Suzuki-Miyaura reactions of aryl sulfonates / halides with arylboronic acids
  • Synthesis of an acromelic acid A analog based 11C-labeled PET tracer
  • Synthesis of (-)-Exiguolide and its analogues
  • Allylation of zinc enolates

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adrian Tlahuext-Aca et al.
Dalton transactions (Cambridge, England : 2003), 43(42), 15997-16005 (2014-09-19)
Ni(0)-catalyzed dehydrogenation of benzylic-type imines was performed to yield asymmetrical tetra-substituted imidazoles and 2-imidazolines. This was achieved with a single operational step while maintaining good selectivity and atom economy. The catalytic system shows low to moderate tolerance for fluoro-, trifluoromethyl-
Sebastijan Kovačič et al.
Macromolecular rapid communications, 36(17), 1605-1611 (2015-07-15)
Metal-organic frameworks (MOFs) nanoparticles in combination with a nonionic surfactant (Pluronic L-121) are used to stabilize dicyclopentadiene (DCPD)-in-water high internal phase emulsions (HIPEs). The resulting HIPEs containing the MIL-100(Fe) nanoparticles (MIL: Materials of Institut Lavoisier) at the interface between the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service