Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

591971

Sigma-Aldrich

Poly(2,2,3,3,4,4,4-heptafluorobutyl methacrylate)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C8H7F7O2)n
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.23

form

solid

refractive index

n20/D 1.383 (lit.)

transition temp

Tg (DSC) 65 °C

solubility

DMSO: soluble
NMP: soluble
THF: soluble
methylene chloride: slightly soluble

Application

Low refractive index polymer for cladding layer in optical waveguides. Transparent, low surface tension polymer.
Poly(2,2,3,3,4,4,4-heptafluorobutyl methacrylate) is a fluorinated polymer used to develop low refractive index optical materials and various optoelectronics including organic photovoltaics, organic field-effect transistors (OFETs), and polymer light-emitting diodes because of its high mechanical strength, high thermal stability, chemical inertness, low dielectric constants, and low water absorptivity.
It is also used to prepare poly[2-(N-carbazolyl)ethyl methacrylate]-block-poly[(2,2,3,3,4,4,4-heptafluorobutyl methacrylate)] (CbzEMAx-b-HFBMAy) diblock copolymers by reversible addition-fragmentation chain transfer (RAFT) polymerization. The availability of a low-surface-energy fluorinated block and an electro-active carbazole fragment in diblock copolymers confers crucial properties for the fabrication of high-quality and high-performance functional thin films.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Well-defined fluoro-and carbazole-containing diblock copolymers: synthesis, characterization and immobilization onto Au-coated silicon surfaces
Savva I, et al.
Royal Society of Chemistry Advances, 2(23), 8741-8751 (2012)
Tailoring and patterning of dielectric interfaces for the development of advanced organic field-effect transistors
Kim Min-Hoi, et al.
Liq. Cryst., 41(3), 310-319 (2014)

Articles

Polymeric materials, with unique refractive index properties, find applications in photonics and optical coatings for various devices.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service