Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

591335

Sigma-Aldrich

PAMAM-OS-trimethoxysilyl dendrimer, generation 0 solution

10 wt. % in isopropanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=0);dendri PAMAM((NHCH2CH2CO2(CH2)3Si(OCH3)3)8
Molecular Weight:
2391.25
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

description

trimethoxysilyl surface group

form

liquid

mol wt

generation 0

feature

core type ethylenediamine core (2-carbon core)

concentration

10 wt. % in isopropanol

no. Surface Groups

8

refractive index

n20/D 1.3859

bp

82.4 °C

density

0.819 g/mL at 25 °C

SMILES string

CO[Si](CCCOC(=O)CCN(CCNC(=O)CCN(CCCN(CCC(=O)NCCN(CCC(=O)OCCC[Si](OC)(OC)OC)CCC(=O)OCCC[Si](OC)(OC)OC)CCC(=O)NCCN(CCC(=O)OCCC[Si](OC)(OC)OC)CCC(=O)OCCC[Si](OC)(OC)OC)CCC(=O)NCCN(CCC(=O)OCCC[Si](OC)(OC)OC)CCC(=O)OCCC[Si](OC)(OC)OC)CCC(=O)OCCC[Si](OC)(OC)OC)(OC)OC

Features and Benefits

Contains 8 surface organosilicon groups.

Legal Information

Manufactured by Dendritech®, Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service