Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

591262

Sigma-Aldrich

Cyclohexylmethyl isothiocyanate

97%

Synonym(s):

(Cyclohexylmethyl) isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H11CH2NCS
CAS Number:
Molecular Weight:
155.26
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5340 (lit.)

bp

222-223 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

S=C=NCC1CCCCC1

InChI

1S/C8H13NS/c10-7-9-6-8-4-2-1-3-5-8/h8H,1-6H2

InChI key

RYBJNWIQNLZNPE-UHFFFAOYSA-N

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service