Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

591173

Sigma-Aldrich

4-Oxo-2,2,6,6-tetramethylpiperidine-1-15N,d17

97 atom %, 98% (CP)

Synonym(s):

2,2,6,6-Tetra(methyl-d3)-4-piperidinone-1,3,3,5,5-d5-1-15N

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9D1715NO
CAS Number:
Molecular Weight:
173.34
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

description

TEMPONE precursor

isotopic purity

97 atom %

Assay

98% (CP)

form

solid

mp

34-38 °C (lit.)

mass shift

M+18

SMILES string

[2H][15N]1C(C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])C(=O)C([2H])([2H])C1(C([2H])([2H])[2H])C([2H])([2H])[2H]

InChI

1S/C9H17NO/c1-8(2)5-7(11)6-9(3,4)10-8/h10H,5-6H2,1-4H3/i1D3,2D3,3D3,4D3,5D2,6D2,10+1/hD

InChI key

JWUXJYZVKZKLTJ-AWFCJHQLSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 2

Flash Point(F)

163.9 °F - closed cup

Flash Point(C)

73.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service