Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

590320

Sigma-Aldrich

Triphenylphosphine(2,5-norbornadiene)rhodium(I) tetrafluoroborate, polymer-bound Fibre-cat®

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

product line

FibreCat®

Quality Level

form

solid

reaction suitability

core: rhodium
reaction type: solution phase peptide synthesis
reagent type: catalyst

InChI

1S/C18H15P.C7H8.BF4.Rh/c1-4-10-16(11-5-1)19(17-12-6-2-7-13-17)18-14-8-3-9-15-18;1-2-7-4-3-6(1)5-7;2-1(3,4)5;/h1-15H;1-4,6-7H,5H2;;/q;;-1;+1

InChI key

QMBLFRMSOHVUJG-UHFFFAOYSA-N

Application

Used as a recyclable supported catalyst for the selective hydrogenation of polyolefins. This resin catalyst exhibits similar selectivity to its homogeneous counterpart.

Features and Benefits

This resin catalyst exhibits similar selectivity to its homogeneous counterpart. Rhodium leaching is very low.

Legal Information

FibreCat is a registered trademark of Johnson Matthey PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service