Skip to Content
MilliporeSigma
All Photos(1)

Documents

589284

Sigma-Aldrich

1-[(Trimethylsilyl)methyl]benzotriazole

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15N3Si
CAS Number:
Molecular Weight:
205.33
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

reaction suitability

reaction type: C-C Bond Formation

mp

56-60 °C (lit.)

SMILES string

C[Si](C)(C)Cn1nnc2ccccc12

InChI

1S/C10H15N3Si/c1-14(2,3)8-13-10-7-5-4-6-9(10)11-12-13/h4-7H,8H2,1-3H3

InChI key

VRGRHLJMRVGQCS-UHFFFAOYSA-N

Application

Reactant for:



  • Alkylation reactions
  • Preparation of inhibitor of p53-MDM2 complex formation
  • Used as a one-carbon synthon for one-carbon homologation of carboxylic acids

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service