Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58925

Sigma-Aldrich

(−)-Isolongifolol

Synonym(s):

(1S,11S)-3,3,7-Trimethyltricyclo[5.3.1.02.8]undecane-11-methanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H26O
CAS Number:
Molecular Weight:
222.37
Beilstein:
3195912
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

optical activity

[α]20/D −58±1°, c = 1% in petroleum ether

mp

113-114 °C

SMILES string

[H][C@@]12CC[C@]3([H])[C@@]1([H])C(C)(C)CCC[C@@]3(C)[C@H]2CO

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service