Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

587788

Sigma-Aldrich

1,6-Diaminohexane-2,2,5,5-d4

98 atom % D

Synonym(s):

1,6-Hexane-2,2,5,5-d4-diamine, Hexamethylene-2,2,5,5-d4-diamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CD2(CH2)2CD2CH2NH2
CAS Number:
Molecular Weight:
120.23
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

form

solid

bp

204-205 °C (lit.)

mp

42-45 °C (lit.)

mass shift

M+4

SMILES string

[2H]C([2H])(N)CCCCC([2H])([2H])N

InChI

1S/C6H16N2/c7-5-3-1-2-4-6-8/h1-8H2/i3D2,4D2

InChI key

NAQMVNRVTILPCV-KHORGVISSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service