Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

586579

Sigma-Aldrich

5-Fluoro-2-oxindole

97%

Synonym(s):

5-Fluoro-1,3-dihydroindol-2-one, 5-Fluoro-2,3-dihydro-2-oxoindole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H6FNO
CAS Number:
Molecular Weight:
151.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

143-147 °C (lit.)

SMILES string

Fc1ccc2NC(=O)Cc2c1

InChI

1S/C8H6FNO/c9-6-1-2-7-5(3-6)4-8(11)10-7/h1-3H,4H2,(H,10,11)

InChI key

DDIIYGHHUMKDGI-UHFFFAOYSA-N

Application

Reactant for preparation of:
  • Glycogen synthase kinase 3 beta (GSK-3β) inhibitors
  • Antitumor agents
  • Cyclooxygenase 1/2 (COX-1/2) and 5-lipoxygenase (5-LOX) inhibitors
  • Multitargeted Receptor Tyrosine Kinase Inhibitors
  • Non-peptide thrombopoietin (TPO) receptor ligands
  • Aurora kinase inhibitors
  • Inhibitors of cyclin-dependent kinase CDK2/cyclin A
  • Selective norepinephrine reuptake inhibitors
  • B-cell lymphoma-extra large molecule (Bcl-XL) inhibitor Z36
  • Inducers of NAD(P)H-quinone oxidoreductase 1 (NQO1)

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service