Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

586277

Sigma-Aldrich

DL-Aspartic acid-1,4-13C2

99 atom % 13C

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HO213CCH2CH(NH2)13CO2H
CAS Number:
Molecular Weight:
135.09
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

isotopic purity

99 atom % 13C

form

solid

mp

>300 °C (dec.) (lit.)

mass shift

M+2

SMILES string

NC(C[13C](O)=O)[13C](O)=O

InChI

1S/C4H7NO4/c5-2(4(8)9)1-3(6)7/h2H,1,5H2,(H,6,7)(H,8,9)/i3+1,4+1

InChI key

CKLJMWTZIZZHCS-CQDYUVAPSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service