Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

58245

Sigma-Aldrich

Isatin-5-sulfonic acid sodium salt dihydrate

≥98.0%

Synonym(s):

Sodium isatin-5-sulfonate dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H4NNaO5S · 2H2O
CAS Number:
Molecular Weight:
285.21
Beilstein:
8398337
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

solid

composition

carbon content, 33.0-34.4%
sulfur, 11.0-11.5 ICP

impurities

6-13% water

mp

≥300 °C (lit.)

functional group

ketone
sulfonic acid

SMILES string

O.O.[Na+].[O-]S(=O)(=O)c1ccc2NC(=O)C(=O)c2c1

InChI

1S/C8H5NO5S.Na.2H2O/c10-7-5-3-4(15(12,13)14)1-2-6(5)9-8(7)11;;;/h1-3H,(H,9,10,11)(H,12,13,14);;2*1H2/q;+1;;/p-1

InChI key

QOAFLTFHMZZJFP-UHFFFAOYSA-M

Application

  • Reactant for preparation of Orally Active Oxytocin Receptor Antagonists

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service