Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

575933

Sigma-Aldrich

Barium periodate

≥99.99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ba(IO4)2
CAS Number:
Molecular Weight:
519.13
EC Number:
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

≥99.99%

reaction suitability

reagent type: oxidant

SMILES string

[Ba++].[O-]I(=O)(=O)=O.[O-]I(=O)(=O)=O

InChI

1S/Ba.2HIO4/c;2*2-1(3,4)5/h;2*(H,2,3,4,5)/q+2;;/p-2

InChI key

SSTZWBSEDRHYPU-UHFFFAOYSA-L

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service