Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

575720

Sigma-Aldrich

Strontium selenide

≥99.99% trace metals basis

Synonym(s):

Strontium monosulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrSe
CAS Number:
Molecular Weight:
166.58
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

≥99.99% trace metals basis

form

powder

mp

1600 °C (lit.)

SMILES string

[Se]=[Sr]

InChI

1S/Se.Sr

InChI key

PFNOMMSIZHKPBO-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

I Marie et al.
Autoimmunity reviews, 16(3), 223-230 (2017-02-01)
This case control study assessed: 1) the relationship of systemic sclerosis (SSc) related to exposure to heavy metals; and 2) the risk of SSc related to occupational exposure in male and female patients. From 2005 to 2008, 100 patients with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service