Skip to Content
MilliporeSigma
All Photos(1)

Documents

575410

Sigma-Aldrich

Poly[(m-phenylenevinylene)-alt-(2,5-dihexyloxy-p-phenylenevinylene)]

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352103

form

solid

composition

trans, 95%

solubility

chloroform: soluble 2%
THF: soluble
toluene: soluble

fluorescence

λex 404 nm; λem 451 nm in chloroform

General description

Poly[(m-phenylenevinylene)-alt-(2,5-dihexyloxy-p-phenylenevinylene)] is a host material that can be used as a blue fluorescent dye in organic electronics.

Application

Poly[(m-phenylenevinylene)-alt-(2,5-dihexyloxy-p-phenylenevinylene)] (PMPDHPV) is a conducting polymer that can be used in the fabrication of organic light emitting diodes (OLEDs). It can also be used as a dye in 3D surface enhanced Raman scattering (SERS) imaging.
Semi-conjugated light-emitting polymer.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fabrication of organic light-emitting diodes using photosynthetic pigments extracted from spinach
Ohtani N, et al.
Japanese Journal of Applied Physics, 50(1S2), 01BC08-01BC08 (2011)
Multimodal delivery of irinotecan from microparticles with two distinct compartments
Rahmani S, et al.
Journal of Controlled Release : Official Journal of the Controlled Release Society, 172(1), 239-245 (2013)
Spatial Analysis of Metal-PLGA Hybrid Microstructures Using 3D SERS Imaging
Strozyk MS, et al.
Advances in Functional Materials, 27(33), 1701626-1701626 (2017)

Articles

LEPs enable a wide range of important applications including sensors, flexible LED displays and lighting devices, optical pump lasers, and potentially polymer diode lasers.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service