Skip to Content
MilliporeSigma
All Photos(2)

Documents

574740

Sigma-Aldrich

trans-Diamminetetrachloroplatinum(IV)

≥99.9% trace metals basis

Synonym(s):

trans- Tetrachlorodiammineplatinum

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Pt(NH3)2Cl4
CAS Number:
Molecular Weight:
370.95
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥99.9% trace metals basis

form

powder

mp

>300 °C (lit.)

SMILES string

N.N.Cl[Pt](Cl)(Cl)Cl

InChI

1S/4ClH.2H3N.Pt/h4*1H;2*1H3;/q;;;;;;+4/p-4

InChI key

RBHOYUZKIANPPG-UHFFFAOYSA-J

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Defais et al.
Chemico-biological interactions, 74(3), 343-352 (1990-01-01)
DNA strand breaks were observed in bacteria treated with Pt(IV) but not Pt(II) antitumor compounds by two methods. First, compounds which cause DNA strand breaks produced an SOS induction signal which was detected by a rapid bacterial assay. In addition
A C Plooy et al.
Cancer research, 44(5), 2043-2051 (1984-05-01)
Several effects of four diamminechloroplatinum compounds (II and IV) in Chinese hamster ovary cells were studied. The two cis-compounds [cis-diamminedichloroplatinum(II) and cis-diamminetetrachloroplatinum(IV)] are known to possess antitumor activity, whereas the two trans-stereoisomers [trans-diamminedichloroplatinum(II) and trans-diamminetetrachloroplatinum(IV)] are inactive. When the effects
H Razaka et al.
Chemico-biological interactions, 60(2), 207-215 (1986-11-01)
After exposure of bacteria to equal concentrations of cis-diamminedichloroplatinum(II) (DDP) and cis-diamminetetrachloroplatinum(IV) (DTP), the intracellular concentration of DTP was an order of magnitude greater than DDP. However, at identical intracellular drug concentrations, the Pt(IV) compound formed only half as many

Articles

Plasmonic nanoparticles have unique optical properties that can be tailored to suit a variety of applications in the biotechnology1–8 and electronics9–16 industries.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service