Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

571172

Sigma-Aldrich

[4-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide solution

0.25 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4CH2NC4H8MgBr
CAS Number:
Molecular Weight:
264.44
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

reaction suitability

reaction type: Grignard Reaction

concentration

0.25 M in THF

bp

65 °C

density

0.916 g/mL at 25 °C

storage temp.

2-8°C

SMILES string

Br[Mg]c1ccc(CN2CCCC2)cc1

InChI

1S/C11H14N.BrH.Mg/c1-2-6-11(7-3-1)10-12-8-4-5-9-12;;/h2-3,6-7H,4-5,8-10H2;1H;/q;;+1/p-1

InChI key

SMCUOBSDAGTZFC-UHFFFAOYSA-M

Application

[4-(1-Pyrrolidinylmethyl)phenyl]magnesium bromide can be used:
  • In the combinatorial preparation of rosamine derivatives, which are applicable as fluorescent probes in monitoring cellular glutathione (GSH) levels.[1]
  • In the synthesis of 4,5-disubstituted-2-aminoimidazole−triazole derivatives having antibiofilm activity.[2]

Legal Information

Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of 4, 5-Disubstituted-2-Aminoimidazole--Triazole Conjugates for Antibiofilm/Antibiotic Resensitization Activity Against MRSA and Acinetobacter baumannii
Su Z, et al.
ChemMedChem, 6(12), 2243-2251 (2011)
Combinatorial rosamine library and application to in vivo glutathione probe
Ahn Y, et al.
Journal of the American Chemical Society, 129(15), 4510-4511 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service