Skip to Content
MilliporeSigma
All Photos(1)

Documents

570818

Sigma-Aldrich

3-Fluoro-2-methylphenylmagnesium chloride solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H3(CH3)MgCl
CAS Number:
Molecular Weight:
168.88
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

bp

65-67 °C (lit.)

density

0.938 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Cc1c(F)cccc1[Mg]Cl

InChI

1S/C7H6F.ClH.Mg/c1-6-4-2-3-5-7(6)8;;/h2-3,5H,1H3;1H;/q;;+1/p-1

InChI key

XVXJKZMOSQDWMQ-UHFFFAOYSA-M

Legal Information

Product of Rieke® Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.0 °F - closed cup

Flash Point(C)

-17.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service