Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

569969

Sigma-Aldrich

Bis(4-aminophenyl)ether-d12

97 atom % D, 97% (CP)

Synonym(s):

4,4′-Oxydianiline-d12

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O(C6D4ND2)2
CAS Number:
Molecular Weight:
212.31
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

97 atom % D

Assay

97% (CP)

mp

188-192 °C (lit.)

mass shift

M+12

SMILES string

[2H]N([2H])c1c([2H])c([2H])c(Oc2c([2H])c([2H])c(N([2H])[2H])c([2H])c2[2H])c([2H])c1[2H]

InChI

1S/C12H12N2O/c13-9-1-5-11(6-2-9)15-12-7-3-10(14)4-8-12/h1-8H,13-14H2/i1D,2D,3D,4D,5D,6D,7D,8D/hD4

InChI key

HLBLWEWZXPIGSM-AZLBOOTHSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Muta. 1B - Repr. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

426.2 °F - closed cup

Flash Point(C)

219 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service