Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

569763

Sigma-Aldrich

(5S)-(−)-2,2,3-Trimethyl-5-benzyl-4-imidazolidinone monohydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H18N2O · HCl
CAS Number:
Molecular Weight:
254.76
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

optical activity

[α]20/D -67°, c = 1 in H2O

mp

157-161 °C (lit.)

functional group

phenyl

SMILES string

Cl.CN1C(=O)[C@H](Cc2ccccc2)NC1(C)C

InChI

1S/C13H18N2O.ClH/c1-13(2)14-11(12(16)15(13)3)9-10-7-5-4-6-8-10;/h4-8,11,14H,9H2,1-3H3;1H/t11-;/m0./s1

InChI key

YIYFEXGDFJLJGM-MERQFXBCSA-N

Application

Metal-free OrganoCatalyst technology for asymmetric catalysis. Catalyzes asymmetric Diels-Alder reactions, 1,3-dipolar additions and pyrrole alkylations in high enantiomeric excess.
Used in first highly enantioselective organocatalytic Diels-Alder reaction[1] and 1,3-dipolar addition.[2]

Legal Information

U.S. Pat. 6,369,243 and related patents apply. For research purposes only.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jen, W. S. et al
Journal of the American Chemical Society, 122, 9874-9874 (2000)
Ahrendt, K. A. et al
Journal of the American Chemical Society, 122, 4243-4243 (2000)
New strategies in organic catalysis: the first enantioselective organocatalytic Friedel-Crafts alkylation.
N A Paras et al.
Journal of the American Chemical Society, 123(18), 4370-4371 (2001-07-18)

Articles

Discover Professor David MacMillan's groundbreaking metal-free asymmetric catalysis using imidazolidinone-based organocatalysts for versatile transformations.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service