Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

566950

Sigma-Aldrich

4-Bromobenzyl mercaptan

97%

Synonym(s):

(4-Bromophenyl)methanethiol, 4-Bromo-α-toluenethiol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2SH
CAS Number:
Molecular Weight:
203.10
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

27-31 °C (lit.)

SMILES string

SCc1ccc(Br)cc1

InChI

1S/C7H7BrS/c8-7-3-1-6(5-9)2-4-7/h1-4,9H,5H2

InChI key

CUCKXDPCCYHFMQ-UHFFFAOYSA-N

Application

4-Bromobenzyl mercaptan may be used to synthesize 4-bromotoluene via desulfurization with molybdenum hexacarbonyl [Mo(CO)6] in tetrahydrofuran.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhengyu Zhang et al.
Chemical research in toxicology, 33(7), 1761-1769 (2020-06-10)
Dioscorea bulbifera L. (DBL), a traditional Chinese medicine, is a well-known herb with hepatotoxicity, and the biochemical mechanisms of the toxic action remain unknown. Diosbulbin B (DSB), a major component of DBL, can induce severer liver injury which requires cytochrome
?Transition-metal promoted reactions. 12. Molybdenum hexacarbonyl-promoted reductive cleavage of the carbon-sulfur bond?
Luh YT and Wong SC
The Journal of Organic Chemistry, 50(25), 5413-5415 (1985)
Franziska Leipoldt et al.
Nature communications, 8(1), 1965-1965 (2017-12-08)
Metalloproteinase inhibitors often feature hydroxamate moieties to facilitate the chelation of metal ions in the catalytic center of target enzymes. Actinonin and matlystatins are  potent metalloproteinase inhibitors that comprise rare N-hydroxy-2-pentyl-succinamic acid warheads. Here we report the identification and characterization of their biosynthetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service