Skip to Content
MilliporeSigma
All Photos(1)

Documents

562335

Sigma-Aldrich

N-Boc-indole-2-boronic acid

Synonym(s):

1-(tert-butoxycarbonyl)indole-2-boronic acid, N-(tert-butoxycarbonyl)indole-2-boronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16BNO4
CAS Number:
Molecular Weight:
261.08
MDL number:
UNSPSC Code:
12352103

mp

85-90 °C (lit.)

SMILES string

CC(C)(C)OC(=O)n1c(cc2ccccc12)B(O)O

InChI

1S/C13H16BNO4/c1-13(2,3)19-12(16)15-10-7-5-4-6-9(10)8-11(15)14(17)18/h4-8,17-18H,1-3H3

InChI key

SVIBPSNFXYUOFT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant involved in:
  • Suzuki-Miyaura cross-coupling reactions
  • Copper-catalyzed trifluoromethylation
  • Palladium-catalyzed benzylation
  • Homocoupling reactions

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service