Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

560669

Sigma-Aldrich

2-Methoxy-5-(3′,7′-dimethyloctyloxy)benzene-1,4-diacetonitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30N2O2
CAS Number:
Molecular Weight:
342.48
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

97%

mp

86.7-88.4 °C (lit.)

SMILES string

COc1cc(CC#N)c(OCCC(C)CCCC(C)C)cc1CC#N

InChI

1S/C21H30N2O2/c1-16(2)6-5-7-17(3)10-13-25-21-15-18(8-11-22)20(24-4)14-19(21)9-12-23/h14-17H,5-10,13H2,1-4H3

InChI key

QRSBGAFJARYUJE-UHFFFAOYSA-N

General description

Monomeric precursor for the light emitting polymer OC1C10-CN-PPV

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service