Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

560502

Sigma-Aldrich

3-(4-Fluorophenyl)propionic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4CH2CH2CO2H
CAS Number:
Molecular Weight:
168.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

86-91 °C (lit.)

functional group

carboxylic acid
fluoro

SMILES string

OC(=O)CCc1ccc(F)cc1

InChI

1S/C9H9FO2/c10-8-4-1-7(2-5-8)3-6-9(11)12/h1-2,4-5H,3,6H2,(H,11,12)

InChI key

ZMKXWDPUXLPHCA-UHFFFAOYSA-N

Application

3-(4-Fluorophenyl)propionic acid may be used in the synthesis of 2-oxopiperazine guanidine analog.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and synthesis of potent and selective 1, 3, 4-trisubstituted-2-oxopiperazine based melanocortin-4 receptor agonists.
Tian X, et al.
Bioorganic & Medicinal Chemistry Letters, 16(17), 4668-4673 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service