Skip to Content
MilliporeSigma
All Photos(1)

Documents

560472

Sigma-Aldrich

2-(4-Morpholino)ethyl isothiocyanate

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H12N2OS
CAS Number:
Molecular Weight:
172.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.5380 (lit.)

bp

163 °C/10 mmHg (lit.)

density

1.129 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

S=C=NCCN1CCOCC1

InChI

1S/C7H12N2OS/c11-7-8-1-2-9-3-5-10-6-4-9/h1-6H2

InChI key

SDNSCXCXVQHBGH-UHFFFAOYSA-N

Application

2-(4-Morpholino)ethyl isothiocyanate may be used to synthesize (R,S)-(2-morpholin-4-yl-ethyl)-thiocarbamic acid O-(5-[1,2] dithiolan-3-yl pentyl) ester hydrochloride.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Claude Guillonneau et al.
European journal of medicinal chemistry, 38(1), 1-11 (2003-02-21)
Molecules containing a dithiolane moiety are widely investigated due to their antioxidant properties. The archetypal representative of this class of compounds is lipoic acid and indeed the lipoic acid-dihydrolipoic acid couple is part of the antioxidant defence system of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service