Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

560456

Sigma-Aldrich

2-Piperidinoethyl isothiocyanate

95%

Synonym(s):

1-(2-Isothiocyanatoethyl)-piperidine, 2-(1-Piperidinyl)ethyl isothiocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2S
CAS Number:
Molecular Weight:
170.28
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

95%

form

liquid

refractive index

n20/D 1.5350 (lit.)

bp

104-105 °C (lit.)

density

1.030 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

S=C=NCCN1CCCCC1

InChI

1S/C8H14N2S/c11-8-9-4-7-10-5-2-1-3-6-10/h1-7H2

InChI key

KMTVCPOROYMLTN-UHFFFAOYSA-N

Application

2-Piperidinoethyl isothiocyanate may be used to synthesize 1-(1-benzylpiperidin-4-yl)-3-(2-(piperidin-1-yl)ethyl)thiourea, a bifunctional thiourea based organocatalyst.
Reactant for synthesis of:
  • Urea and thiourea derivatvies as antmalarial chemotherapeutics
  • Neuropeptides S antagonist
  • N-substituted imidazoles
  • Thio-containing compounds

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diversity oriented synthesis of tri-substituted methane containing aminouracil and hydroxynaphthoquinone/hydroxycoumarin moiety using organocatalysed multicomponent reactions in aqueous medium.
Bharti R and Parvin T.
Royal Society of Chemistry Advances, 5(82), 66833-66839 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service