Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

560065

Sigma-Aldrich

9,9-Dihexyl-2,7-dibromofluorene

97%

Synonym(s):

2,7-Dibromo-9,9′-bihexylfluorene, 2,7-Dibromo-9,9-di-n -hexylfluorene, 2,7-Dibromo-9,9-dihexyl-9H -fluorene, 2,7-Dibromo-9,9-dihexylfluorene, 9,9-Dihexyl-2,7-dibromofluorene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H32Br2
CAS Number:
Molecular Weight:
492.33
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

solid

mp

67-71 °C (lit.)

SMILES string

CCCCCCC1(CCCCCC)c2cc(Br)ccc2-c3ccc(Br)cc13

InChI

1S/C25H32Br2/c1-3-5-7-9-15-25(16-10-8-6-4-2)23-17-19(26)11-13-21(23)22-14-12-20(27)18-24(22)25/h11-14,17-18H,3-10,15-16H2,1-2H3

InChI key

OXFFIMLCSVJMHA-UHFFFAOYSA-N

General description

Intermediate for polymeric light-emitting diodes.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service