Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559881

Sigma-Aldrich

2-Butyl-5-chloro-1H-imidazole-4-carboxaldehyde

97%

Synonym(s):

2-Butyl-4-chloro-5-formylimidazole

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3N2HClCHOCH2CH2CH2CH3
CAS Number:
Molecular Weight:
186.64
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

form

solid

mp

97-100 °C (lit.)

SMILES string

CCCCc1nc(C=O)c(Cl)[nH]1

InChI

1S/C8H11ClN2O/c1-2-3-4-7-10-6(5-12)8(9)11-7/h5H,2-4H2,1H3,(H,10,11)

InChI key

JLVIHQCWASNXCK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Butyl-5-chloro-1H-imidazole-4-carboxaldehyde may be used in the preparation of the following symmetrical esters of dialkyl 4-(2-butyl-5-chloro-1H-imidazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate:
  • dimethyl 4-(2-butyl-5-chloro-1H-imidazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
  • diethyl 4-(2-butyl-5-chloro-1H-imidazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
  • diisopropyl 4-(2-butyl-5-chloro-1Himidazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
  • diisobutyl 4-(2-butyl-5-chloro-1H-imidazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
  • di-tert-butyl 4-(2-butyl-5-chloro-1Himidazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate
  • bis(2-methoxyethyl) 4-(2-butyl-5-chloro-1Himidazol-4-yl)-2,6-dimethyl-1,4-dihydropyridine-3,5-dicarboxylate

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry & Biology Interface.
El-Moselhy TF.
Chemistry & Biology, 3(2), 123-136 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service