Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559474

Sigma-Aldrich

3,4,5-Trifluorobenzoyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F3C6H2COCI
CAS Number:
Molecular Weight:
194.54
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.4900 (lit.)

bp

173-174 °C (lit.)

density

1.4850 g/mL at 25 °C (lit.)

SMILES string

Fc1cc(cc(F)c1F)C(Cl)=O

InChI

1S/C7H2ClF3O/c8-7(12)3-1-4(9)6(11)5(10)2-3/h1-2H

InChI key

YRUNCQNKZIQTEO-UHFFFAOYSA-N

Application

3,4,5-Trifluorobenzoyl chloride may be used as one of the reactant to synthesize 3,4,5-trifluoro-N-{2-[4-(2-methoxyphenyl)piperazin-1-yl]ethyl}-N-pyridin-2-ylbenzamide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

[18F]/19F exchange in fluorine containing compounds for potential use in 18F-labelling strategies.
Blom E, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 52(12), 504-511 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service