Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

559423

Sigma-Aldrich

4-Methyl-3-(trifluoromethyl)aniline

98%

Synonym(s):

3-(Trifluoromethyl)-4-methylaniline

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H3(CH3)NH2
CAS Number:
Molecular Weight:
175.15
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.490 (lit.)

bp

204 °C (lit.)

density

1.220 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

Cc1ccc(N)cc1C(F)(F)F

InChI

1S/C8H8F3N/c1-5-2-3-6(12)4-7(5)8(9,10)11/h2-4H,12H2,1H3

InChI key

JBCDCYFEJQHTTA-UHFFFAOYSA-N

Application

4-Methyl-3-(trifluoromethyl)aniline may be used as a starting reagent in the synthesis of tert-butyl 4-methyl-3-(trifluoromethyl)phenylcarbamate.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.1 °F - closed cup

Flash Point(C)

101.7 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis, and biological evaluation of 3-(1 H-1, 2, 3-triazol-1-yl) benzamide derivatives as potent pan Bcr-Abl inhibitors including the threonine315? isoleucine315 mutant.
Li Y, et al.
Journal of Medicinal Chemistry, 55(22), 10033-10046 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service