Skip to Content
MilliporeSigma
All Photos(1)

Documents

558591

Sigma-Aldrich

2,5-Difluoroiodobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3I
CAS Number:
Molecular Weight:
239.99
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5600 (lit.)

bp

178-179 °C (lit.)

density

1.998 g/mL at 25 °C (lit.)

SMILES string

Fc1ccc(F)c(I)c1

InChI

1S/C6H3F2I/c7-4-1-2-5(8)6(9)3-4/h1-3H

InChI key

WBYVNDUCUMNZPM-UHFFFAOYSA-N

Application

2,5-Difluoroiodobenzene (1,4-Difluoro-2-iodobenzene) may be used in the synthesis of 3,6-difluoro-1,2-phenylenediboronic acid and 3,6-difluoro-2-iodobenzoic acid.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

185.0 °F - closed cup

Flash Point(C)

85 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Basicity Gradient-Driven Migration of Iodine: Conferring Regioflexibility on the Substitution of Fluoroarenes.
Rausis T and Schlosser M.
European Journal of Organic Chemistry, 19, 3351-3358 (2002)
Influence of Fluorination and Boronic Group Synergy on the Acidity and Structural Behavior of o-Phenylenediboronic Acids.
Durka K, et al.
Organometallics, 33(7), 1608-1616 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service