Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

558400

Sigma-Aldrich

1-(3-Chloro-4-fluorophenyl)biguanide hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)NHCNHNHCNHNH2 · xHCl
Molecular Weight:
229.64 (free base basis)
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
To inquire about this product 558400, please contact your local MilliporeSigma office or dealer. Contact Technical Service

Assay

97%

mp

209-213 °C (lit.)

SMILES string

Cl.NC(=N)NC(=N)Nc1ccc(F)c(Cl)c1

InChI

1S/C8H9ClFN5.ClH/c9-5-3-4(1-2-6(5)10)14-8(13)15-7(11)12;/h1-3H,(H6,11,12,13,14,15);1H

InChI key

DLLQSZUCBVOQDX-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gabriela Cabral et al.
Developmental cell, 50(3), 355-366 (2019-07-16)
Centrosomes, the predominant sites of microtubule nucleation and anchorage, coordinate spindle assembly and cell division in animal cells. At the onset of mitosis, centrioles accumulate microtubule-organizing pericentriolar material (PCM) in a process termed centrosome maturation. To what extent centrosome maturation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service