Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

558001

Sigma-Aldrich

2-Chlorothioanisole

96%

Synonym(s):

2-Chlorophenyl methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4SCH3
CAS Number:
Molecular Weight:
158.65
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

refractive index

n20/D 1.608 (lit.)

bp

239-240 °C (lit.)

functional group

chloro
thioether

SMILES string

CSc1ccccc1Cl

InChI

1S/C7H7ClS/c1-9-7-5-3-2-4-6(7)8/h2-5H,1H3

InChI key

IHLDFHCSSCVPQW-UHFFFAOYSA-N

General description

2-Chlorothioanisole can be prepared from the reaction between N,N-dimethylformamide, 1,2-dichlorobenzene, finely ground KOH and methyl mercaptan.[1] Benzene dipole moment value analysis of 2-chlorothioanisole indicates that it exists predominantly in trans- form.[2] 1H NMR spectrum of 2-chlorothioanisole has been studied in acetone-d6 solution.[3]

Application

2-Chlorothioanisole may be used to synthesize 2-chlorophenyl methyl sulfone.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

158.0 °F - closed cup

Flash Point(C)

70 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The preferred conformations of 2, 4-dichlorophenol, 2, 4-dichloroanisole and their sulphur analogues.
Lumbroso H, et al.
Journal of Molecular Structure, 43(1), 87-95 (1978)
Development of the Large-Scale Preparation of 2-(Methanesulfonyl) benzenesulfonyl Chloride.
Meckler H and Herr RJ.
Organic Process Research & Development, 16(4), 550-555 (2012)
Analysis of chlorophenylmethylsulfides in the urine of rats injected with chlorobenzene by high performance liquid chromatography.
M Yoshida et al.
Industrial health, 23(4), 283-287 (1985-01-01)
The proximate coupling constant, 5 J (H, CH3), and the torsional mobility of the thiomethyl group in some thioanisole derivatives.
Schaefer T, et al.
Canadian Journal of Chemistry, 69(4), 620-624 (1991)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service