Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

557234

Sigma-Aldrich

5-Chloro-2-fluorophenylboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(F)B(OH)2
CAS Number:
Molecular Weight:
174.37
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.21

form

solid

mp

122-127 °C (lit.)

functional group

chloro
fluoro

SMILES string

OB(O)c1cc(Cl)ccc1F

InChI

1S/C6H5BClFO2/c8-4-1-2-6(9)5(3-4)7(10)11/h1-3,10-11H

InChI key

GGTUVWGMCFXUAS-UHFFFAOYSA-N

Application

Reactant involved in:
  • Suzuki cross-coupling reactions
  • Synthesis of biaryl amides with muscarinic acetylcholine receptor subtype M1 agonistic activity
  • Synthesis of kinesin spindle protein inhibitors
  • GABA α2/3 agonist preparation

Other Notes

Contains varying amounts of anhydride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service