Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

556785

Sigma-Aldrich

5-Amino-2-[(4-aminophenyl)amino]benzenesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4NHC6H3(NH2)SO3H
CAS Number:
Molecular Weight:
279.31
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

mp

244 °C (lit.)

SMILES string

Nc1ccc(Nc2ccc(N)cc2S(O)(=O)=O)cc1

InChI

1S/C12H13N3O3S/c13-8-1-4-10(5-2-8)15-11-6-3-9(14)7-12(11)19(16,17)18/h1-7,15H,13-14H2,(H,16,17,18)

InChI key

VKURVCNKVWKGLX-UHFFFAOYSA-N

Application

5-Amino-2-[(4-aminophenyl)amino]benzenesulfonic acid may be used to synthesize unsymmetrical bis-azo dyes.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis Castedo, J. Chen, Charles Chiu, S. Collins, Donald Craig
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 31a (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service