55617
N-(Hydroxymethyl)benzamide
≥98.0%
Synonym(s):
Benzamidomethanol
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C8H9NO2
CAS Number:
Molecular Weight:
151.16
Beilstein:
2045129
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
≥98.0%
mp
95-100 °C
SMILES string
OCNC(=O)c1ccccc1
InChI
1S/C8H9NO2/c10-6-9-8(11)7-4-2-1-3-5-7/h1-5,10H,6H2,(H,9,11)
InChI key
UOUBPDZUBVJZOQ-UHFFFAOYSA-N
Other Notes
Aminomethylating reagent; employed e.g. for preparing α-amino phosphonic acids; Preparation of benzamidomethyl protected prodrugs which are fairly stable in physiological medium
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M. Sting et al.
Synthesis, 132-132 (1990)
W J Tenn et al.
Organic letters, 3(1), 75-78 (2001-06-30)
[figure: see text] The rate constants for the hydronium ion, hydroxide, and water-catalyzed breakdown of N-(hydroxymethyl)benzamide (1), 4-chloro-N-(hydroxymethyl)benzamide (2), and 2,4-dichloro-N-(hydroxymethyl)benzamide (3) in H2O, at 25 degrees C, I = 1.0 (KCl), have been determined. The reactions of 1, 2
J.J. Getz et al.
The Journal of Organic Chemistry, 57, 1702-1702 (1992)
D Ross et al.
Life sciences, 32(6), 597-604 (1983-02-07)
N-Formylbenzamide and benzamide were characterised by high pressure liquid chromatography and mass spectrometry as products of the metabolism of N-hydroxymethylbenzamide in incubation mixtures with mouse liver preparations and isolated hepatocytes. This biotransformation occurred predominantly in 9000g and microsomal supernatant fractions
R J Simmonds et al.
Anti-cancer drug design, 3(2), 91-101 (1988-08-01)
Of the three N-hydroxymethyl compounds in the title, the first two have pronounced antineoplastic activity while the latter is biologically inactive. Crystals of the triazene have monoclinic symmetry with a = 8.540(1), b = 6.346(4), c = 22.460(5)A, beta =
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service