Skip to Content
MilliporeSigma
All Photos(1)

Documents

556025

Sigma-Aldrich

tert-Butyl methyl succinate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CO2CCH2CH2CO2CH3
CAS Number:
Molecular Weight:
188.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.421 (lit.)

bp

85-95 °C/20 mmHg (lit.)

density

1.022 g/mL at 25 °C (lit.)

SMILES string

COC(=O)CCC(=O)OC(C)(C)C

InChI

1S/C9H16O4/c1-9(2,3)13-8(11)6-5-7(10)12-4/h5-6H2,1-4H3

InChI key

KGLNZUDBQPORRI-UHFFFAOYSA-N

General description

tert-Butyl methyl succinate is an ester. Mixture of its threo- and erythro- isomeric forms were obtained by the addition reaction between tert-butyl alcohol and 4-oxo-but-3-enoate in the presence of lithium tert-butoxide (catalyst).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danheiser RL.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 689-689 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service