555827
2,4-Dicyano-3-ethyl-3-methylglutarimide
97%
Synonym(s):
4-Ethyl-4-methyl-2,6-dioxo-3,5-piperidine dicarbonitrile
About This Item
Assay
97%
form
solid
mp
183-187 °C (lit.)
SMILES string
CCC1(C)C(C#N)C(=O)NC(=O)C1C#N
InChI
1S/C10H11N3O2/c1-3-10(2)6(4-11)8(14)13-9(15)7(10)5-12/h6-7H,3H2,1-2H3,(H,13,14,15)
InChI key
ISARNQWKABTJEY-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service