Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

555614

Sigma-Aldrich

2,8-Dichloroquinoline

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H5Cl2N
CAS Number:
Molecular Weight:
198.05
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

105-108 °C (lit.)

SMILES string

Clc1ccc2cccc(Cl)c2n1

InChI

1S/C9H5Cl2N/c10-7-3-1-2-6-4-5-8(11)12-9(6)7/h1-5H

InChI key

VAXOCTXTVIVOQE-UHFFFAOYSA-N

General description

2,8-Dichloroquinoline can be synthesized from the reaction between 8-chloroquinoline-N-oxide and phosphoryl trichloride.

Application

2,8-Dichloroquinoline may be used in the neat synthesis of 8-chloro-2-(thiophen-2-ylsulfanyl)quinolone. It may also be used in the preparation of 2-methyl-4-[-2′-(8′-chloroquinolyl)]-but-3-yn-2-ol.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of n-chloroquinolines and n-ethynylquinolines (n= 2, 4, 8): homo and heterocoupling reactions.
Rodriguez JG, et al.
Tetrahedron, 61(38), 9042-9051 (2005)
Catalyst-Free Preparation of Heterocyclic Thienyl Sulfides.
Barrett K, et al.
Synthetic Communications, 45(24), 2857-2860 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service