Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

555363

Sigma-Aldrich

Lithium deuteride

98 atom % D

Synonym(s):

Lithium hydride-d

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiD
CAS Number:
Molecular Weight:
8.96
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.12

isotopic purity

98 atom % D

mass shift

M+1

SMILES string

[Li][2H]

InChI

1S/Li.H/i;1+1

InChI key

SIAPCJWMELPYOE-IEOVAKBOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium deuteride (LiD) is a stable isotopic compound that is commonly used in deuterium storage and energy storage applications because of its high melting point, low density, high deuterium concentration, and exceptional thermal stability in both vacuum and inert gases. It can also be used as a potential raw material for the synthesis of organic deuterides, deuterated solvents, and deuterated polymers.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - Water-react 1

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Effect of air humidity on microstructure and phase composition of lithium deuteride corrosion products
L Xiaobo and L Jiping
Corrosion Science, 115, 129-134 (2017)
Lattice dynamics of lithium hydride and lithium deuteride: effect of long-range three-body forces
Laplaze D
Journal of Physics C: Solid State Physics, 10(18), 3499-3499 (1977)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service