Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

555282

Sigma-Aldrich

Cesium chloride/sodium chloride (2:1) eutectic

AnhydroBeads, 99.99% trace metals basis

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Cl3Cs2Na
CAS Number:
Molecular Weight:
395.16
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.99% trace metals basis

impurities

≤200.0 ppm Trace Metal Analysis

mp

497-500 °C (lit.)

SMILES string

[Na+].[Cl-].[Cl-].[Cl-].[Cs+].[Cs+]

InChI

1S/3ClH.2Cs.Na/h3*1H;;;/q;;;3*+1/p-3

InChI key

OLSYZSZVAMPEGY-UHFFFAOYSA-K

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service