Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

554111

Sigma-Aldrich

2,3,4-(Trimethoxy)bromobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H2(OCH3)3
CAS Number:
Molecular Weight:
247.09
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

refractive index

n20/D 1.549 (lit.)

bp

105-110 °C/2 mmHg (lit.)

density

1.291 g/mL at 25 °C (lit.)

functional group

bromo

storage temp.

2-8°C

SMILES string

COc1ccc(Br)c(OC)c1OC

InChI

1S/C9H11BrO3/c1-11-7-5-4-6(10)8(12-2)9(7)13-3/h4-5H,1-3H3

InChI key

HKCQJLVIFMFOTG-UHFFFAOYSA-N

General description

2,3,4-(Trimethoxy)bromobenzene can be prepared from 1,2,3-(trimethoxy)benzene via bromination.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemo-and Regioselective Bromination of Aromatic Compounds in the Presence of ?-picolinium Bromochromate (?-PBC)/CH.
Mahmoodi NO, et al.
Orient. J. Chem., 27(2), 429-434 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service