Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

554006

Sigma-Aldrich

Tris(cyclopentadienyl)terbium(III)

99.9% trace metals basis

Synonym(s):

TbCp3

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C5H5)3Tb
CAS Number:
Molecular Weight:
354.20
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

Assay

99.9% trace metals basis

form

solid

reaction suitability

core: terbium
reagent type: catalyst

mp

235 °C (dec.) (lit.)

SMILES string

[Tb].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2.[CH]3[CH][CH][CH][CH]3

InChI

1S/3C5H5.Tb/c3*1-2-4-5-3-1;/h3*1-5H;

InChI key

FMNCAPFXHVPBJH-UHFFFAOYSA-N

General description

Atomic number of base material: 65 Terbium

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Sol. 2 - Water-react 2

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service