Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

552437

Sigma-Aldrich

2-Fluoro-5-iodobenzoic acid

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H3FICOOH
CAS Number:
Molecular Weight:
266.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

164-168 °C (lit.)

SMILES string

OC(=O)c1cc(I)ccc1F

InChI

1S/C7H4FIO2/c8-6-2-1-4(9)3-5(6)7(10)11/h1-3H,(H,10,11)

InChI key

QNNJHBNTHVHALE-UHFFFAOYSA-N

Application

2-Fluoro-5-iodobenzoic acid may be used in the preparation of 1-N-ethyl 6-iodoquinolonic acid and 1-N-cyclopropyl 6-iodoquinolonic acid.[1] It may also be used as a precursor for the generation of electron transfer dissociation (ETD) reagents.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electron-transfer reagent anion formation via electrospray ionization and collision-induced dissociation.
Huang TY, et al.
Analytical Chemistry, 78(21), 7387-7391 (2006)
Synthesis and biological activity of 4 ″-O-acyl derivatives of 14-and 15-membered macrolides linked to ω-quinolone-carboxylic unit.
Skugor MM, et al.
Bioorganic & Medicinal Chemistry, 18(17), 6547-6558 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service